siRNA Details
| Gene Symbol: Xpo4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020506.1 | NP_065252.1 | 7 | 809 | ||
| GenBank mRNA | AF145021.1 | - | 7 | 809 | ||
| AK084907.1 | - | 8 | 994 | |||
| AK129430.1 | - | 7 | 804 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Xpo4| Gene Symbol: Xpo4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020506.1 | NP_065252.1 | 7 | 809 | ||
| GenBank mRNA | AF145021.1 | - | 7 | 809 | ||
| AK084907.1 | - | 8 | 994 | |||
| AK129430.1 | - | 7 | 804 | |||