siRNA Details
Gene Symbol: Cxcl14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019568.2 | NP_062514.2 | 3, 4 | 662 | ||
GenBank mRNA | AB041614.1 | - | 3, 4 | 662 | ||
AF144754.1 | - | 3, 4 | 486 | |||
AF192557.1 | - | 3, 4 | 660 | |||
AF352784.1 | - | 3, 4 | 272 | |||
AF352785.1 | - | 3, 4 | 272 | |||
AK004615.1 | - | 3, 4 | 506 | |||
AK014351.1 | - | 3, 4 | 452 | |||
AK076112.1 | - | 3, 4 | 504 | |||
AK155356.1 | - | 3, 4 | 500 | |||
BC147274.1 | - | 3, 4 | 406 | 9056321 | ||
BC147275.1 | - | 3, 4 | 406 | 9056329 |
