siRNA Details
| Gene Symbol: Bcam | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020486.2 | NP_065232.1 | 8 | 1052 | ||
| GenBank mRNA | AB035511.1 | - | 8 | 1013 | ||
| AF109160.1 | - | 8 | 1052 | |||
| AF221507.1 | - | 8 | 1002 | |||
| AF346663.1 | - | 8 | 990 | |||
| AK040564.1 | - | 8 | 1036 | |||
| AK054001.1 | - | 8 | 1033 | |||
| AK075704.1 | - | 8 | 1017 | |||
| AK158439.1 | - | 8 | 1018 | |||
| BC004826.1 | - | 8 | 1036 | 2616148 | ||

Bcam
Genomic Map