siRNA Details
| Gene Symbol: Slc25a20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020520.4 | NP_065266.1 | 8, 9 | 981 | ||
| GenBank mRNA | AK075624.1 | - | 8, 9 | 982 | ||
| AK152436.1 | - | 8, 9 | 952 | |||
| AK177588.1 | - | 1, 2 | 29 | |||
| AK183069.1 | - | 1, 2 | 29 | |||
| AK211911.1 | - | 1, 2 | 29 | |||
| BC052871.1 | - | 8, 9 | 904 | 30032324 | ||

Slc25a20
Genomic Map