siRNA Details
| Gene Symbol: Smpdl3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020561.2 | NP_065586.3 | 1 | 162 | ||
| GenBank mRNA | AK002455.1 | - | 1 | 101 | ||
| AK151683.1 | - | 1 | 48 | |||
| AK152282.1 | - | 1 | 63 | |||
| AK194237.1 | - | 1 | 70 | |||
| BC139288.1 | - | 1 | 54 | 8862310 | ||
| BC139289.1 | - | 1 | 54 | 8862311 | ||
| Y08135.1 | - | 1 | 116 | |||

Smpdl3a
Genomic Map