siRNA Details
Gene Symbol: Smpdl3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020561.2 | NP_065586.3 | 7 | 1084 | ||
GenBank mRNA | AK002455.1 | - | 7 | 1023 | ||
AK151683.1 | - | 7 | 970 | |||
AK152282.1 | - | 7 | 985 | |||
BC139288.1 | - | 7 | 976 | 8862310 | ||
BC139289.1 | - | 7 | 976 | 8862311 | ||
Y08135.1 | - | 7 | 1038 |
