siRNA Details
| Gene Symbol: Cramp1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020608.3 | NP_065633.2 | 10 | 1278 | ||
| GenBank mRNA | AK089393.1 | - | 10 | 1531 | ||
| AK147411.1 | - | 10 | 1279 | |||
| AK147449.1 | - | 10 | 1279 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cramp1l| Gene Symbol: Cramp1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020608.3 | NP_065633.2 | 10 | 1278 | ||
| GenBank mRNA | AK089393.1 | - | 10 | 1531 | ||
| AK147411.1 | - | 10 | 1279 | |||
| AK147449.1 | - | 10 | 1279 | |||