siRNA Details
Gene Symbol: Gmnn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020567.2 | NP_065592.1 | 3 | 204 | ||
GenBank mRNA | AF068780.1 | - | 3 | 351 | ||
AK131959.1 | - | 3 | 178 | |||
BC020061.1 | - | 3 | 356 | 3988065 |
