siRNA Details
| Gene Symbol: Noc2l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021303.2 | NP_067278.2 | 6 | 733 | ||
| GenBank mRNA | AK076287.1 | - | 5 | 775 | ||
| AK078560.1 | - | 6 | 709 | |||
| AK133834.1 | - | 6 | 733 | |||
| AK157765.1 | - | 6 | 725 | |||
| AK164071.1 | - | 2 | 1650 | |||
| AK184541.1 | - | 2 | 177 | |||
| BC020013.1 | - | 6 | 701 | 3663482 | ||

Noc2l
Genomic Map