siRNA Details
| Gene Symbol: Sec61a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021305.3 | NP_067280.1 | 10 | 1231 | ||
| GenBank mRNA | AF145254.1 | - | 10 | 1167 | ||
| AF222748.1 | - | 10 | 1168 | |||
| AK017615.1 | - | 9 | 1015 | |||
| AK077701.1 | - | 10 | 1231 | |||
| AK160602.1 | - | 10 | 1215 | |||
| BC005458.1 | - | 10 | 1174 | 3494001 | ||

Sec61a2
Genomic Map