siRNA Details
| Gene Symbol: Jmy | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021310.3 | NP_067285.2 | 8 | 2518 | ||
| GenBank mRNA | AB221690.1 | - | 8 | 2016 | ||
| AF201390.1 | - | 8 | 2142 | |||
| BC090835.1 | - | 8 | 2399 | 5705427 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Jmy| Gene Symbol: Jmy | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021310.3 | NP_067285.2 | 8 | 2518 | ||
| GenBank mRNA | AB221690.1 | - | 8 | 2016 | ||
| AF201390.1 | - | 8 | 2142 | |||
| BC090835.1 | - | 8 | 2399 | 5705427 | ||