siRNA Details
| Gene Symbol: Jmy | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021310.3 | NP_067285.2 | 1, 2 | 1517 | ||
| GenBank mRNA | AB221690.1 | - | 1, 2 | 1015 | ||
| AF201390.1 | - | 1, 2 | 1141 | |||
| AK139934.1 | - | 1, 2 | 1146 | |||
| AK141957.1 | - | 1, 2 | 1518 | |||
| BC020052.1 | - | 1, 2 | 1442 | |||
| BC069906.1 | - | 1, 2 | 1371 | |||
| BC090835.1 | - | 1, 2 | 1398 | 5705427 | ||

Jmy
Genomic Map