siRNA Details
| Gene Symbol: Adck2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178873.3 | NP_849204.1 | 5 | 1872 | ||
| GenBank mRNA | AK028557.1 | - | 5 | 1876 | ||
| AK028568.1 | - | 5 | 1873 | |||
| AK161225.1 | - | 5 | 1875 | |||
| BC069944.1 | - | 5 | 1620 | 30446117 | ||

Adck2
Genomic Map