siRNA Details
Gene Symbol: Hacd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021345.2 | NP_067320.2 | 4 | 505 | ||
GenBank mRNA | AK050556.1 | - | 4 | 499 | ||
AK051735.1 | - | 4 | 464 | |||
AK052865.1 | - | 4 | 502 | |||
AK159494.1 | - | 4 | 471 | |||
BC031755.1 | - | 4 | 489 | 4488391 | ||
BC057023.1 | - | 4 | 481 | 6813640 | ||
Z97207.2 | - | 4 | 331 |
