siRNA Details
Gene Symbol: Hacd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021345.2 | NP_067320.2 | 7, 8 | 835 | ||
GenBank mRNA | AK050556.1 | - | 7, 8 | 829 | ||
AK051735.1 | - | 7, 8 | 794 | |||
AK052865.1 | - | 7, 8 | 832 | |||
AK159494.1 | - | 7, 8 | 801 | |||
BC031755.1 | - | 7, 8 | 819 | 4488391 | ||
BC057023.1 | - | 7, 8 | 811 | 6813640 |
