siRNA Details
| Gene Symbol: Zfp318 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021346.2 | NP_067321.2 | 4 | 1396 | ||
| NM_207671.4 | NP_997554.2 | 4 | 1396 | |||
| GenBank mRNA | AF159455.1 | - | 3 | 805 | ||
| AF227194.1 | - | 3 | 805 | |||
| BC150730.1 | - | 3 | 727 | 9087641 | ||

Zfp318
Genomic Map