siRNA Details
| Gene Symbol: Hic2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178922.3 | NP_849253.2 | 3 | 1526 | ||
| GenBank mRNA | AF117382.1 | - | 1 | 250 | ||
| AJ868292.1 | - | 2 | 1490 | |||
| AK028357.1 | - | 2 | 1338 | |||
| AK084975.1 | - | 3 | 1449 | |||
| AK129266.1 | - | 3 | 1512 | |||
| AK145386.1 | - | 1 | 633 | |||
| AK156332.1 | - | 3 | 1542 | |||
| AK157276.1 | - | 3 | 1542 | |||
| BC065124.1 | - | 1525 | 30537019 | |||

Hic2
Genomic Map