siRNA Details
| Gene Symbol: Hic2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178922.3 | NP_849253.2 | 3 | 1729 | ||
| GenBank mRNA | AF117382.1 | - | 1 | 453 | ||
| AJ868292.1 | - | 2 | 1693 | |||
| AK028357.1 | - | 2 | 1541 | |||
| AK084975.1 | - | 3 | 1652 | |||
| AK129266.1 | - | 3 | 1715 | |||
| AK145386.1 | - | 1 | 836 | |||
| AK156332.1 | - | 3 | 1745 | |||
| AK157276.1 | - | 3 | 1745 | |||
| BC065124.1 | - | 1728 | 30537019 | |||

Hic2
Genomic Map