siRNA Details
| Gene Symbol: Sh3kbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135727.2 | NP_001129199.1 | 12 | 1587 | ||
| NM_001135728.2 | NP_001129200.1 | 7 | 589 | |||
| NM_021389.6 | NP_067364.2 | 11 | 1152 | |||
| GenBank mRNA | AK004636.1 | - | 11 | 1152 | ||
| AK028299.1 | - | 7 | 593 | |||
| AK030839.1 | - | 11 | 1153 | |||
| AK038540.1 | - | 14 | 1451 | |||
| AK049182.1 | - | 12 | 1588 | |||
| AK080930.1 | - | 12 | 1452 | |||
| AK161735.1 | - | 7 | 670 | |||
| AK172022.1 | - | 12 | 1568 | |||
| AK213880.1 | - | 3 | 153 | |||
| BC034646.1 | - | 11 | 1156 | |||
| BC052863.1 | - | 11 | 1097 | |||

Sh3kbp1
Genomic Map