siRNA Details
Gene Symbol: Zbtb32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021397.2 | NP_067372.2 | 2 | 847 | ||
GenBank mRNA | AF241232.1 | - | 1 | 756 | ||
BC137687.1 | - | 1 | 750 | 8860706 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Zbtb32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021397.2 | NP_067372.2 | 2 | 847 | ||
GenBank mRNA | AF241232.1 | - | 1 | 756 | ||
BC137687.1 | - | 1 | 750 | 8860706 |