siRNA Details
Gene Symbol: Zbtb32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021397.2 | NP_067372.2 | 5 | 1218 | ||
GenBank mRNA | AF241232.1 | - | 4 | 1127 | ||
AK015881.1 | - | 3 | 301 | |||
BC137687.1 | - | 4 | 1121 | 8860706 | ||
BC144837.1 | - | 4 | 883 |
