siRNA Details
| Gene Symbol: Slc43a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021398.3 | NP_067373.2 | 5 | 777 | ||
| GenBank mRNA | AF188622.1 | - | 5 | 771 | ||
| AK086962.1 | - | 4 | 1206 | |||
| BC089603.1 | - | 4 | 477 | 6758493 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc43a3| Gene Symbol: Slc43a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021398.3 | NP_067373.2 | 5 | 777 | ||
| GenBank mRNA | AF188622.1 | - | 5 | 771 | ||
| AK086962.1 | - | 4 | 1206 | |||
| BC089603.1 | - | 4 | 477 | 6758493 | ||