siRNA Details
| Gene Symbol: Trem1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021406.5 | NP_067381.1 | 3 | 613 | ||
| GenBank mRNA | AF241219.1 | - | 3 | 568 | ||
| AK089439.1 | - | 3 | 582 | |||
| AK171753.1 | - | 3 | 582 | |||
| BC111875.1 | - | 2 | 225 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Trem1| Gene Symbol: Trem1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021406.5 | NP_067381.1 | 3 | 613 | ||
| GenBank mRNA | AF241219.1 | - | 3 | 568 | ||
| AK089439.1 | - | 3 | 582 | |||
| AK171753.1 | - | 3 | 582 | |||
| BC111875.1 | - | 2 | 225 | |||