siRNA Details
| Gene Symbol: Fam184b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021416.3 | NP_067391.3 | 10 | 2170 | ||
| GenBank mRNA | AB093291.1 | - | 9 | 1699 | ||
| AB257856.1 | - | 10 | 2283 | |||
| AK081567.1 | - | 10 | 1959 | |||
| BC056379.1 | - | 10 | 1977 | 6850901 | ||

Fam184b
Genomic Map