siRNA Details
Gene Symbol: Stx6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021433.3 | NP_067408.1 | 7 | 847 | ||
GenBank mRNA | AF237814.1 | - | 7 | 657 | ||
AK009690.1 | - | 7 | 802 | |||
AK018167.1 | - | 7 | 835 | |||
AK019106.1 | - | 7 | 809 | |||
AK031673.1 | - | 7 | 826 | |||
AK044905.1 | - | 7 | 808 | |||
AK049124.1 | - | 7 | 807 | |||
AK151549.1 | - | 7 | 801 | |||
AK163167.1 | - | 220 | ||||
BC029205.1 | - | 7 | 764 | 4039560 | ||
CN456515.1 | - | 3 | 227 |
