siRNA Details
| Gene Symbol: Stx6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021433.3 | NP_067408.1 | 5 | 596 | ||
| GenBank mRNA | AF237814.1 | - | 5 | 406 | ||
| AK009690.1 | - | 5 | 551 | |||
| AK018167.1 | - | 5 | 584 | |||
| AK019106.1 | - | 5 | 558 | |||
| AK031673.1 | - | 5 | 575 | |||
| AK044905.1 | - | 5 | 557 | |||
| AK049124.1 | - | 5 | 556 | |||
| AK151549.1 | - | 5 | 550 | |||
| AK159392.1 | - | 5 | 568 | |||
| BC029205.1 | - | 5 | 513 | 4039560 | ||
| BQ749205.1 | - | 5 | 569 | |||

Stx6
Genomic Map