siRNA Details
| Gene Symbol: Pga5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021453.4 | NP_067428.2 | 4, 5 | 539 | ||
| GenBank mRNA | AF240776.1 | - | 4, 5 | 448 | ||
| AK004109.1 | - | 4, 5 | 478 | |||
| AK053965.1 | - | 4, 5 | 540 | |||
| AK168924.1 | - | 4, 5 | 478 | |||
| BC119523.2 | - | 4, 5 | 497 | 40056066 | ||

Pga5
Genomic Map