siRNA Details
| Gene Symbol: Slco1c1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177772.1 | NP_001171243.1 | 9 | 1546 | ||
| NM_021471.2 | NP_067446.1 | 9 | 1546 | |||
| GenBank mRNA | AK045782.1 | - | 1549 | |||
| AK048926.1 | - | 9 | 1516 | |||
| AK161010.1 | - | 1516 | ||||
| AY007379.1 | - | 9 | 1382 | |||
| BC078456.1 | - | 9 | 1496 | 30360663 | ||

Slco1c1
Genomic Map