siRNA Details
Gene Symbol: Slco1c1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177772.1 | NP_001171243.1 | 3 | 529 | ||
NM_021471.2 | NP_067446.1 | 3 | 529 | |||
GenBank mRNA | AK045782.1 | - | 532 | |||
AK048926.1 | - | 3 | 499 | |||
AK161010.1 | - | 499 | ||||
AY007379.1 | - | 3 | 365 | |||
BC078456.1 | - | 3 | 479 | 30360663 |
