siRNA Details
| Gene Symbol: Rnase4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021472.4 | NP_067447.2 | 2 | 556 | ||
| NM_201239.3 | NP_957691.1 | 2 | 853 | |||
| GenBank mRNA | AB041044.1 | - | 2 | 462 | ||
| AK050451.1 | - | 2 | 453 | |||
| AY762362.1 | - | 2 | 455 | |||
| BC005569.1 | - | 2 | 430 | 3967265 | ||
| BC061105.1 | - | 2 | 437 | 30296907 | ||
| BC066843.1 | - | 2 | 517 | 30059308 | ||

Rnase4
Genomic Map