siRNA Details
Gene Symbol: Treh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277847.1 | NP_001264776.1 | 5, 6 | 527 | ||
NM_001277851.1 | NP_001264780.1 | 5, 6 | 531 | |||
NM_021481.3 | NP_067456.1 | 6, 7 | 632 | |||
GenBank mRNA | AF136944.1 | - | 6, 7 | 627 | ||
AK002566.1 | - | 5, 6 | 530 | |||
AK008912.1 | - | 6, 7 | 617 | |||
BC019214.1 | - | 5, 6 | 522 | 5064708 |
