siRNA Details
| Gene Symbol: Pex5l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021483.4 | NP_067458.2 | 2 | 324 | ||
| GenBank mRNA | AB032591.1 | - | 2 | 319 | ||
| HQ646191.1 | - | 1 | 1028 | |||
| HQ646192.1 | - | 2 | 442 | |||
| HQ646193.1 | - | 2 | 348 | |||
| HQ646194.1 | - | 2 | 348 | |||

Pex5l
Genomic Map