siRNA Details
| Gene Symbol: Pex5l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289505.1 | NP_001276434.1 | 2, 3 | 238 | ||
| NM_021483.4 | NP_067458.2 | 2, 3 | 370 | |||
| GenBank mRNA | AB032591.1 | - | 2, 3 | 365 | ||
| AK039764.1 | - | 2, 3 | 238 | |||
| BY129053.1 | - | 2, 3 | 238 | |||
| HQ646191.1 | - | 1, 2 | 1074 | |||
| HQ646192.1 | - | 2, 3 | 488 | |||
| HQ646193.1 | - | 2, 3 | 394 | |||
| HQ646194.1 | - | 2, 3 | 394 | |||
| HQ646195.1 | - | 2, 3 | 262 | |||
| HQ646196.1 | - | 2, 3 | 262 | |||

Pex5l
Genomic Map