siRNA Details
| Gene Symbol: Hibadh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145567.1 | NP_663542.1 | 8 | 975 | ||
| GenBank mRNA | AB051067.1 | - | 8 | 1033 | ||
| AK078175.1 | - | 8 | 926 | |||
| BC003914.1 | - | 8 | 975 | 3484897 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hibadh| Gene Symbol: Hibadh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145567.1 | NP_663542.1 | 8 | 975 | ||
| GenBank mRNA | AB051067.1 | - | 8 | 1033 | ||
| AK078175.1 | - | 8 | 926 | |||
| BC003914.1 | - | 8 | 975 | 3484897 | ||