siRNA Details
| Gene Symbol: Sumf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145937.3 | NP_666049.2 | 2 | 308 | ||
| GenBank mRNA | AK151874.1 | - | 2 | 309 | ||
| AK159192.1 | - | 2 | 297 | |||
| AK160917.1 | - | 2 | 290 | |||
| AK161203.1 | - | 2 | 301 | |||
| BC026981.1 | - | 2 | 281 | 5365580 | ||
| BY331659.1 | - | 2 | 308 | |||

Sumf1
Genomic Map