siRNA Details
| Gene Symbol: Pvrl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021495.4 | NP_067470.1 | 3 | 821 | ||
| NM_021496.3 | NP_067471.1 | 3 | 821 | |||
| NM_021497.2 | NP_067472.1 | 3 | 821 | |||
| GenBank mRNA | AF195833.1 | - | 3 | 818 | ||
| AF195834.1 | - | 3 | 628 | |||
| AF195835.1 | - | 3 | 628 | |||
| BC125588.1 | - | 3 | 747 | 40130103 | ||
| BC132187.1 | - | 3 | 747 | 40130464 | ||
| BU701648.1 | - | 2 | 187 | |||
| CJ102084.1 | - | 2 | 401 | |||

Pvrl3
Genomic Map