siRNA Details
| Gene Symbol: Pvrl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021495.4 | NP_067470.1 | 5 | 1139 | ||
| NM_021496.3 | NP_067471.1 | 5 | 1139 | |||
| NM_021497.2 | NP_067472.1 | 5 | 1139 | |||
| GenBank mRNA | AF195833.1 | - | 5 | 1136 | ||
| AF195834.1 | - | 5 | 946 | |||
| AF195835.1 | - | 5 | 946 | |||
| AK011949.1 | - | 5 | 1139 | |||
| AK013863.1 | - | 224 | ||||
| BC125588.1 | - | 5 | 1065 | 40130103 | ||
| BC132187.1 | - | 5 | 1065 | 40130464 | ||
| BU701648.1 | - | 4 | 505 | |||

Pvrl3
Genomic Map