siRNA Details
| Gene Symbol: Maea | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021500.2 | NP_067475.2 | 5 | 599 | ||
| GenBank mRNA | AF263247.1 | - | 5 | 561 | ||
| AK007307.1 | - | 5 | 570 | |||
| AK049424.1 | - | 5 | 619 | |||
| AK075637.1 | - | 4 | 439 | |||
| AK088854.1 | - | 5 | 607 | |||
| AK145800.1 | - | 5 | 662 | |||
| AK152523.1 | - | 5 | 607 | |||
| AK168788.1 | - | 5 | 625 | |||
| AK169149.1 | - | 5 | 607 | |||
| BC039054.1 | - | 5 | 585 | 2648667 | ||
| BC058687.1 | - | 5 | 597 | 6411707 | ||

Maea
Genomic Map