siRNA Details
| Gene Symbol: Nup160 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021512.2 | NP_067487.1 | 14 | 2187 | ||
| GenBank mRNA | AF104415.2 | - | 14 | 2187 | ||
| AK155492.1 | - | 13 | 1561 | |||
| AK164802.1 | - | 13 | 1540 | |||
| AK171218.1 | - | 13 | 1573 | |||
| BC052450.1 | - | 13 | 1551 | 6409240 | ||
| BC054523.1 | - | 13 | 1551 | 6409240 | ||

Nup160
Genomic Map