siRNA Details
Gene Symbol: Nup160 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021512.2 | NP_067487.1 | 8 | 1578 | ||
GenBank mRNA | AF104415.2 | - | 8 | 1578 | ||
AK050952.1 | - | 4 | 306 | |||
AK155492.1 | - | 7 | 952 | |||
AK160685.1 | - | 7 | 940 | |||
AK164802.1 | - | 7 | 931 | |||
AK171218.1 | - | 7 | 964 | |||
BC052450.1 | - | 7 | 942 | 6409240 | ||
BC054523.1 | - | 7 | 942 | 6409240 |
