siRNA Details
| Gene Symbol: Pdzk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001146001.1 | NP_001139473.1 | 6 | 958 | ||
| NM_021517.2 | NP_067492.2 | 6 | 1004 | |||
| GenBank mRNA | AF220100.1 | - | 6 | 943 | ||
| AF474247.1 | - | 5 | 942 | |||
| AK006269.1 | - | 6 | 1478 | |||
| AK012070.1 | - | 6 | 994 | |||
| AK014879.1 | - | 6 | 1261 | |||
| AK029752.1 | - | 6 | 1462 | |||
| AK029764.1 | - | 6 | 1472 | |||
| AK165415.1 | - | 6 | 939 | |||
| BC013512.1 | - | 5 | 927 | 4234949 | ||

Pdzk1
Genomic Map