siRNA Details
| Gene Symbol: Pdzk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001146001.1 | NP_001139473.1 | 7 | 1071 | ||
| NM_021517.2 | NP_067492.2 | 7 | 1117 | |||
| GenBank mRNA | AF220100.1 | - | 7 | 1056 | ||
| AF474247.1 | - | 6 | 1055 | |||
| AK006269.1 | - | 7 | 1591 | |||
| AK012070.1 | - | 7 | 1107 | |||
| AK014879.1 | - | 7 | 1374 | |||
| AK029752.1 | - | 7 | 1575 | |||
| AK029764.1 | - | 7 | 1585 | |||
| AK165415.1 | - | 7 | 1052 | |||
| BC013512.1 | - | 6 | 1040 | 4234949 | ||

Pdzk1
Genomic Map