siRNA Details
| Gene Symbol: Med12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021521.2 | NP_067496.2 | 32, 33 | 4698 | ||
| GenBank mRNA | AF071310.1 | - | 31, 32 | 4448 | ||
| AK220216.1 | - | 11, 12 | 1396 | |||
| BC057085.1 | - | 5, 6 | 1015 | |||
| BC057119.1 | - | 31, 32 | 4432 | |||
| BC090654.1 | - | 32, 33 | 4696 | |||
| BC110696.1 | - | 2, 3 | 174 | |||
| BC145447.1 | - | 31, 32 | 4383 | |||

Med12
Genomic Map