siRNA Details
| Gene Symbol: Psmd14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021526.2 | NP_067501.2 | 5, 6 | 558 | ||
| GenBank mRNA | AK011974.1 | - | 5, 6 | 507 | ||
| AK012013.1 | - | 5, 6 | 489 | |||
| AK031863.1 | - | 5, 6 | 496 | |||
| AK078134.1 | - | 5, 6 | 510 | |||
| AK150280.1 | - | 5, 6 | 494 | |||
| AK151104.1 | - | 5, 6 | 541 | |||
| AK151733.1 | - | 5, 6 | 541 | |||
| BC003742.1 | - | 5, 6 | 463 | 3599671 | ||
| Y13071.1 | - | 5, 6 | 479 | |||

Psmd14
Genomic Map