siRNA Details
| Gene Symbol: C1galt1c1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021550.3 | NP_067525.1 | 2 | 456 | ||
| GenBank mRNA | AB030184.1 | - | 2 | 368 | ||
| AB091728.1 | - | 1 | 284 | |||
| AK005118.1 | - | 2 | 386 | |||
| AK008040.1 | - | 2 | 403 | |||
| AY159320.1 | - | 2 | 371 | |||
| BC029909.1 | - | 2 | 422 | 3154812 | ||

C1galt1c1
Genomic Map