siRNA Details
| Gene Symbol: Hgh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021555.2 | NP_067530.2 | 2 | 704 | ||
| GenBank mRNA | AB041552.1 | - | 2 | 704 | ||
| AK085815.1 | - | 2 | 698 | |||
| AK194217.1 | - | 2 | 66 | |||
| AK206665.1 | - | 1 | 38 | |||
| BC061251.1 | - | 2 | 668 | 30249656 | ||

Hgh1
Genomic Map