siRNA Details
| Gene Symbol: Evc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021292.2 | NP_067267.2 | 17 | 2566 | ||
| GenBank mRNA | AJ250841.1 | - | 17 | 2546 | ||
| AK039759.1 | - | 17 | 2537 | |||
| AK148262.1 | - | 17 | 2539 | |||
| AK161910.1 | - | 4 | 1216 | |||

Evc
Genomic Map