siRNA Details
| Gene Symbol: Midn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AB036882.1 | - | 5 | 893 | ||
| AK154535.1 | - | 4 | 837 | |||
| BC034719.1 | - | 5 | 863 | 3710931 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Midn| Gene Symbol: Midn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AB036882.1 | - | 5 | 893 | ||
| AK154535.1 | - | 4 | 837 | |||
| BC034719.1 | - | 5 | 863 | 3710931 | ||