siRNA Details
| Gene Symbol: Jph2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001205076.1 | NP_001192005.1 | 6 | 2398 | ||
| NM_021566.2 | NP_067541.1 | 5 | 2478 | |||
| GenBank mRNA | AB024447.1 | - | 5 | 2455 | ||
| AK157145.1 | - | 6 | 2398 | |||
| AY974567.1 | - | 2 | 220 | |||

Jph2
Genomic Map