siRNA Details
| Gene Symbol: Nek7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021605.3 | NP_067618.1 | 10 | 1024 | ||
| GenBank mRNA | AF217650.1 | - | 9 | 875 | ||
| AK035502.1 | - | 10 | 1023 | |||
| AK088173.1 | - | 10 | 1073 | |||
| AK149308.1 | - | 10 | 1072 | |||
| AK165583.1 | - | 8 | 793 | |||
| BC026541.1 | - | 10 | 1014 | |||
| BC037697.1 | - | 10 | 1001 | 3994871 | ||

Nek7
Genomic Map